CO₂ absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Ascarite(II)®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

208085000 208080025 208081000
ACRO208085000EA 297 GBP
ACRO208085000 ACRO208080025 ACRO208081000
CO₂ absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Ascarite(II)®
Desiccators Desiccants
CAS No.: 81133-20-2

MDL No.: MFCD00132264
UN: 1823
Hazard Class: 8
Packing group: II
GHS Symbols: GHS05
Appearance: Brown Granules
Storage Temperature: Ambient
Signal word: Danger
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR