Ammonium thiocyanate 99+%, extra_pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate

206500010 206505000 206500025 206500100
ACRO206500010EA 61.5 GBP
ACRO206500010 ACRO206505000 ACRO206500025 ACRO206500100
Ammonium thiocyanate 99+%, extra_pure
Ammonium thiocyanate
Formula: NH₄SCN
MW: 76.12 g/mol
Melting Pt: 149 °C
Density: 1.3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564

Order Now

Specification Test Results

Appearance White Crystals
Titration Argentometric ≥99.0 % (on dried basis)
Water ≤1.0 %
pH 4.0 to 6.0 (5 % in water)
Iron (Fe) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR