Zirconium tetrachloride, anhydrous 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Zirconium (IV) chloride

206410050 206411000 206415000 206410025
ACRO206410050EA 27.2 GBP
ACRO206410050 ACRO206411000 ACRO206415000 ACRO206410025
Zirconium tetrachloride, anhydrous 98%
Zirconium tetrachloride
Formula: Cl₄Zr
Boiling Pt: 3577 °C (1013 hPa)
Melting Pt: 437 °C (subl.)
Density: 2,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011306
CAS Number: 10026-11-6
EINECS: 233-058-2
UN: 2503
ADR: 8,III
Merck Index: 12,10307

Order Now

Specification Test Results

Appearance White Powder
Assay ≥97.5 % (ZrCl4)
Aluminium (Al) ≤100 ppm
Chromium (Cr) ≤100 ppm
Hafnium (Hf) ≤100 ppm
Iron (Fe) ≤1000 ppm
Silicon (Si) ≤150 ppm
Titanium (Ti) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR