D-(-)-Lyxose mixture of anomers 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

205230050 205230250
ACRO205230050EA 45.9 GBP
ACRO205230050 ACRO205230250
D-(-)-Lyxose mixture of anomers 99+%
D(-)-Lyxose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00064362
CAS Number: 1114-34-7
EINECS: 214-212-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥99.0 % (ELSD)
Specific optical rotation -13° to -15° (20°C, 589 nm) (c=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR