Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Sodium isethionate, Sodium hydroxyethyl sulphonate, 2-Hydroxyethanesulphonic acid sodium salt

205000010 205005000
ACRO205000010EA 62.8 GBP
ACRO205000010 ACRO205005000
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate) 98%
Isethionic acid sodium salt (Sodium 2-hydroxyethanesulphonate)
Formula: C₂H₅NaO₄S
MW: 148.11 g/mol
Melting Pt: 191…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00007534
CAS Number: 1562-00-1
EINECS: 216-343-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 191 to 195 °C
Titration after Ion exchange ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR