Hydroxy naphthol blue disodium salt ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Hydroxynaphthol blue sodium salt, Hydroxy napthol blue, Sodium (E)-3-hydroxy-4-((2-hydroxy-6-sulpho-3-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate

204880050 204881000 204885000
ACRO204880050EA 24.8 GBP
ACRO204880050 ACRO204881000 ACRO204885000
Hydroxy naphthol blue disodium salt ACS
Hydroxy naphthol blue disodium salt

CAS No.: 165660-27-5

Formula: C₂₀H₁₂N₂O₁₁S₃·2Na
Melting Pt: 274 °C
Storage Temperature: Ambient
MDL Number: MFCD00004075
CAS Number: 165660-27-5

Order Now

Specification Test Results

Appearance Blue to dark purple Crystalline powder or crystals
Infrared spectrum Conforms
Suitability (for Ca determination) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR