Hexylamine 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1-Hexylamine, 1-Aminohexane

204720050 204725000 204721000 204720010
ACRO204720050EA 21.5 GBP
ACRO204720050 ACRO204725000 ACRO204721000 ACRO204720010
Hexylamine 99%
Hexylamine
Formula: H₃C(CH₂)₄CH₂NH₂
MW: 101.19 g/mol
Boiling Pt: 131
Melting Pt: –19 °C
Density: 0.75 g/cm³
Flash Pt: 27
Storage Temperature: Ambient
MDL Number: MFCD00008240
CAS Number: 111-26-2
EINECS: 203-851-8
UN: 2734
ADR: 8 I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4165 to 1.4195 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR