Phosphorous acid 98%, extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

201150020 201155000 201150010 201150100
ACRO201150020EA 101 GBP
ACRO201150020 ACRO201155000 ACRO201150010 ACRO201150100
Phosphorous acid 98%, extra pure
Phosphorous acid
Formula: H₃O₃P
Melting Pt: 73 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00137258
CAS Number: 13598-36-2
EINECS: 237-066-7
UN: 2834
ADR: 8,III

Order Now

Specification Test Results

Appearance White Crystals
Titration with NaOH 97.5 to 102.5 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR