Diphosphorus pentoxide 98%, extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

200890010 200890025 200890100 200892500
ACRO200890010EA 60.3 GBP
ACRO200890010 ACRO200890025 ACRO200890100 ACRO200892500
Diphosphorus pentoxide 98%, extra pure
di-Phosphorus pentoxide
Formula: P₂O₅
Melting Pt: 340…360 °C (subl.)
Density: 2,253 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011440
CAS Number: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II
Merck Index: 13,07441

Order Now

Specification Test Results

Appearance White Hygroscopic crystalline powder
Titration with NaOH ≥97.5 %
Reducing substance (P2O3) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR