Ammonium bromide 99+% for analysis

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

199980010 199980250 199982500
ACRO199980010EA 40.7 GBP
ACRO199980010 ACRO199980250 ACRO199982500
Ammonium bromide 99+% for analysis
Ammonium bromide
Formula: H₄BrN
Boiling Pt: 235 °C (760 mmHg)
Melting Pt: 452 °C (subl.)
Density: 2,42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011419
CAS Number: 12124-97-9
EINECS: 235-183-8
Merck Index: 13,00505

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥99.0 % (on dry substance)
Loss on drying ≤0.2 % (105°C)
Sulfated ash ≤0.05 %
pH 4.5 to 6.0 (5 % solution)
Chloride (Cl) ≤0.2 %
Bromate (BrO3) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Barium (Ba) ≤20 ppm
Copper (Cu) ≤2 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤2 ppm
Nickel (Ni) ≤2 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR