Palladium hydroxide (20% Pd) on carbon (Pearlman's catalyst) wetted with ca. 60% water, powder, unreduced

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

199620100 199620500
ACRO199620100EA 602 GBP
ACRO199620100 ACRO199620500
Palladium hydroxide (20% Pd) on carbon (Pearlman's catalyst) wetted with ca. 60% water, powder, unreduced
Palladium hydroxide

CAS No.: 12135-22-7


Appearance: Black Powder

Formula: H₂O₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00064599
CAS Number: 12135-22-7

Order Now

Specification Test Results

Appearance Black Powder
Water 45.0 to 55.0 %
Palladium (Pd) 18.0 to 22.0 % On dry basis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR