Copper(II) sulphate, anhydrous 98+% analysis_for

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Cupric sulphate

197711000 197715000 197710025
ACRO197711000EA 23.6 GBP
ACRO197711000 ACRO197715000 ACRO197710025
Copper(II) sulphate, anhydrous 98+% analysis_for
Copper (II) sulphate
Formula: CuSO₄
MW: 159.61 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3077
ADR: 9 III
Merck Index: 12,02722

Order Now

Specification Test Results

Appearance Light green to grey Powder
Titration Iodometric ≥98.0 %
Loss on drying ≤1 % (300°C)
Chloride (Cl) ≤100 ppm
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Magnesium (Mg) ≤50 ppm
Nickel (Ni) ≤50 ppm
Zinc (Zn) ≤20 ppm
Total nitrogen (as N) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR