Molecular sieve 13X (1.0 nm, 10 Å) 4 - 8 mesh

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Linde 13X

197300050 197305000
ACRO197300050EA 173 GBP
ACRO197300050 ACRO197305000
Molecular sieve 13X (1.0 nm, 10 Å) 4 - 8 mesh
Molecular sieve 13X (1.0 nm, 10 Å)
MDL Number: MFCD00164506
CAS Number: 63231-69-6

Order Now

Specification Test Results

Appearance Beige to light brown Beads
Particle size 4 to 8 mesh
Bulk density ≥600 g/l
Residual water ≤1.5 % (550°C, 2 h)
CO2 adsorption capacity ≥11.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR