Silver tetrafluoroborate 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Argentous tetrafluoroborate

197090100 197090500 197090010
ACRO197090100EA 119 GBP
ACRO197090100 ACRO197090500 ACRO197090010
Silver tetrafluoroborate 99%
Silver tetrafluoroborate
Formula: AgBF₄
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00003408
CAS Number: 14104-20-2
EINECS: 237-956-5
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance Beige Crystalline powder
Assay ≥43.92 % (BF4)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR