Sodium thiocyanate 98%, extra_pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Sodium rhodanide

196765000 196760025 196762500
ACRO196765000EA 43.6 GBP
ACRO196765000 ACRO196760025 ACRO196762500
Sodium thiocyanate 98%, extra_pure
Sodium thiocyanate
Formula: NaSCN
MW: 81.07 g/mol
Melting Pt: 287 °C
Storage Temperature: Ambient
MDL Number: MFCD00011123
CAS Number: 540-72-7
EINECS: 208-754-4
Merck Index: 13,09401

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Titration Argentometric ≥97.5 % (on dried substance)
Loss on drying ≤2.5 %
pH 5.5 to 7.5 (5 % at 20°C)
Iron (Fe) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR