Potassium iodide 99%, briquettes, extra_pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Dipotassium diiodide, Kali iodide

196730025 196735000 196730100 196731000
ACRO196730025EA 432 GBP
ACRO196730025 ACRO196735000 ACRO196730100 ACRO196731000
Potassium iodide 99%, briquettes, extra_pure
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C (760 mmHg)
Melting Pt: 680 °C
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Appearance White Briquettes
Titration Argentometric ≥98.5 % (on dried substance)
Loss on drying ≤1 %
Iodate (IO3) ≤4 ppm
Sulfate (SO4) ≤200 ppm
Thiosulphate (S2O3) ≤100 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR