Iodine 99.5%, extra_pure, sublimed

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

196561000 196565000 196560025
ACRO196561000EA 35.1 GBP
ACRO196561000 ACRO196565000 ACRO196560025
Iodine 99.5%, extra_pure, sublimed
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 III
Merck Index: 13,05036

Order Now

Specification Test Results

Appearance Dark grey Metallic spheres
Titration Iodometric ≥99.4 %
Chloride and Bromide ≤0.005 %
Insoluble matter Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR