Potassium tetrahydroborate 98+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potassium tetrahydridoborate, Potassium borohydride

196551000 196555000 196550025 196550050
ACRO196551000EA 65.4 GBP
ACRO196551000 ACRO196555000 ACRO196550025 ACRO196550050
Potassium tetrahydroborate 98+%
Potassium tetrahydroborate
Formula: H₄BK
MW: 53.94 g/mol
Melting Pt: 500
Storage Temperature: Ambient
MDL Number: MFCD00011396
CAS Number: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3 I
Merck Index: 12,07777

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR