Platinum(IV) chloride 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

195400010 195400050 195402500
ACRO195400010EA 140 GBP
ACRO195400010 ACRO195400050 ACRO195402500
Platinum(IV) chloride 99%
Platinum (IV) chloride
Formula: Cl₄Pt
Melting Pt: 370 °C
Density: 4,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011182
CAS Number: 13454-96-1
EINECS: 236-645-1
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance Brown Powder and/or chunks
Trace metal analysis ≤500 ppm (Sum Ru+Rh+Ir+Pd)
Calcium (Ca) ≤250 ppm
Copper (Cu) ≤25 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤25 ppm
Magnesium (Mg) ≤250 ppm
Platinum (Pt) 57.0 to 58.5 %
Silicon (Si) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR