Palladium, black 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

195150010 195150050
ACRO195150010EA 301 GBP
ACRO195150010 ACRO195150050
Palladium, black 98%
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
UN: 3089
ADR: 4.1 II

Order Now

Specification Test Results

Appearance Black Powder
Assay ≥97.5 % (Pd)
Calcium (Ca) ≤25 ppm
Copper (Cu) ≤25 ppm
Gold (Au) ≤50 ppm
Iridium (Ir) ≤50 ppm
Iron (Fe) ≤25 ppm
Lead (Pb) ≤25 ppm
Magnesium (Mg) ≤25 ppm
Platinum (Pt) ≤50 ppm
Rhodium (Rh) ≤50 ppm
Ruthenium (Ru) ≤50 ppm
Silicon (Si) ≤50 ppm
Silver (Ag) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR