Palladium 0,5%, pellets_1/8-inch alumina for use as a catalyst

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

195080500 195082500 195080010
ACRO195080500EA 94.8 GBP
ACRO195080500 ACRO195082500 ACRO195080010
Palladium 0,5%, pellets_1/8-inch alumina for use as a catalyst
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now

Specification Test Results

Appearance Grey Pellets
Assay 0.5 % (Pd)
Loss on drying 1 % (typically)
Additional info loss on abrasion: 3.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR