Zinc 99.995% (trace metals basis), powder

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

194500050 194500500
ACRO194500050EA 70.3 GBP
ACRO194500050 ACRO194500500
Zinc 99.995% (trace metals basis), powder
Zinc
Formula: Zn
Boiling Pt: 907 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3,II

Order Now

Specification Test Results

Appearance Grey Powder
Total trace metal impurities ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR