Aluminium sulphate 99.999% (trace metals basis), extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Aluminum sulphate

192430050 192430250 192431000
ACRO192430050EA 36.7 GBP
ACRO192430050 ACRO192430250 ACRO192431000
Aluminium sulphate 99.999% (trace metals basis), extra pure
Aluminium sulphate
Formula: Al₂O₁₂S₃
Melting Pt: 770 °C
Density: 2,672 g/cm³ (22.5 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003423
CAS Number: 10043-01-3
EINECS: 233-135-0
Merck Index: 12,00381

Order Now

Specification Test Results

Appearance White Powder and/or chunks
Titration Complexometric ≥15 % (Al) Typical
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR