L(+)-Lactic acid 90% 90% in water

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid

189870010 189872500 189870051
ACRO189870010EA 68.2 GBP
ACRO189870010 ACRO189872500 ACRO189870051
L(+)-Lactic acid 90% 90% in water
L(+)-Lactic acid
Formula: C₃H₆O₃
Boiling Pt: 125 °C
Melting Pt: ~ 53 °C
Density: 1.2 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2
Merck Index: 13,05352

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20°C, 589 nm)
Sulfated ash ≤0.1 %
Density (20°C) 1.2 to 1.21 g/ml
Identification Passes test
Titration excess NaOH ≥88 %
Chloride (Cl) ≤0.2 %
Sulfate (SO4) ≤0.25 %
Arsenic (As) ≤3 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Sugars Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR