Tetramethylammonium bromide 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

187631000 187635000
ACRO187631000EA 31.6 GBP
ACRO187631000 ACRO187635000
Tetramethylammonium bromide 98%
Tetramethylammonium bromide
Formula: C₄H₁₂BrN
MW: 154.05 g/mol
Boiling Pt: ∼360 °C (1013 hPa)
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011626
CAS Number: 64-20-0
EINECS: 200-581-2
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Water ≤1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR