Benzotriazole 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole

183881000 183885000 183880025
ACRO183881000EA 33.5 GBP
ACRO183881000 ACRO183885000 ACRO183880025
Benzotriazole 99%
Benzotriazole
Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 204 °C (15 mmHg)
Melting Pt: 94…99 °C
Flash Pt: 170 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005699
CAS Number: 95-14-7
EINECS: 202-394-1
UN: 3077
ADR: 9 III
Merck Index: 13,01109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba54cc42ef5ad9dbae6d7f118a3af1ae642287&vid=ce57b3f0d2583e32220d1e191d0202ad452e6475&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder or crystals or granules or needles or flakes
Infrared spectrum Conforms
Melting point 94 to 99 °C
Titration with NaOH ≥98.5 %
Solubility (2 % in water) Clear colorless solution
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR