Cumene 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Phenylpropane, Isopropylbenzene

181010010 181010025
ACRO181010010EA 31.9 GBP
ACRO181010010 ACRO181010025
Cumene 99%
Cumene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 151
Melting Pt: –96 °C
Density: 0.862 g/cm³
Flash Pt: 31
Storage Temperature: Ambient
MDL Number: MFCD00008881
CAS Number: 98-82-8
EINECS: 202-704-5
UN: 1918
ADR: 3 III
Merck Index: 13,02644

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4930 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR