Ethylene glycol monomethyl ether (2-methoxyethanol) 99+%, extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

180790010 180790025 180790250
ACRO180790010EA 39.9 GBP
ACRO180790010 ACRO180790025 ACRO180790250
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+%, extra pure
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula: C₃H₈O₂
Boiling Pt: 124.6 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.96 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4040 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.0 %
Color scale ≤15 APHA
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR