Petroleum spirit, 40…60 °C ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

180210010 180210025 180210050 180210250
ACRO180210010EA 43.9 GBP
ACRO180210010 ACRO180210025 ACRO180210050 ACRO180210250
Petroleum spirit, 40…60 °C ACS
Petroleum spirit
Boiling Pt: 40…60 °C (1013 hPa)
Density: 0.65 g/cm³
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.01 % (Coulometric)
Boiling point Not less than 90% distils within each range
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity/alkalinity ≤0.0001 meq/g corresp. 0.0006%
Substance darkened by H2SO4 ≤10 APHA
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR