1-Naphthalenemethanol 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Naphthalen-1-ylmethanol

180110020 180110100
ACRO180110020EA 21.6 GBP
ACRO180110020 ACRO180110100
1-Naphthalenemethanol 98%
1-Naphthalenemethanol
Formula: C₁₁H₁₀O
MW: 158.20 g/mol
Boiling Pt: 301 °C (715 mmHg)
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00004044
CAS Number: 4780-79-4
EINECS: 225-324-1

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 59 to 62 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR