2-Iodobenzyl alcohol 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

179700100 179700500
ACRO179700100EA 42.5 GBP
ACRO179700100 ACRO179700500
2-Iodobenzyl alcohol 99%
2-Iodobenzyl alcohol
Formula: C₇H₇IO
Boiling Pt: 145 °C (10 -rr)
Melting Pt: 88…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004610
CAS Number: 5159-41-1
EINECS: 225-933-2

Order Now

Specification Test Results

Appearance White to yellow to pink Powder or needles
Infrared spectrum Conforms
Melting point 88 to 93 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR