Iodosobenzene diacetate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Iodobenzene diacetate, (Diacetoxyiodo)benzene

176560250 176561000 176565000
ACRO176560250EA 54.2 GBP
ACRO176560250 ACRO176561000 ACRO176565000
Iodosobenzene diacetate 98%
Iodosobenzene diacetate
Formula: C₁₀H₁₁IO₄
MW: 322.1 g/mol
Melting Pt: 161…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00008692
CAS Number: 3240-34-4
EINECS: 221-808-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 164 °C
Titration Iodometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR