Syringaldazine

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 4-[[(3,5-dimethoxy-4-oxo-1-cyclohexa-2,5-dienylidene)methylhydrazo]methylidene]-2,6-dimethoxy-1-cyclohexa-2,5-dienone, 4-Hydroxy-3,5-dimethoxybenzaldehyde azine, 3,5-Dimethoxy-4-hydroxybenzalazine, 4,4'-(Hydrazine-1,2-diylidenebis(methanylylidene))bis(2,6-dimethoxyphenol)

175930010 175930050
ACRO175930010EA 67.3 GBP
ACRO175930010 ACRO175930050
Syringaldazine
Syringaldazine

  • Bioactive

Formula: C₁₈H₂₀N₂O₆
Melting Pt: 209…210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008363
CAS Number: 14414-32-5

Order Now

Specification Test Results

Appearance Yellow to green Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 209 to 213 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR