1,3-Cyclopentanedione 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Cyclopentane-1,3-dione

175720050 175720250
ACRO175720050EA 119 GBP
ACRO175720050 ACRO175720250
1,3-Cyclopentanedione 99%
1,3-Cyclopentanedione
Formula: C₅H₆O₂
Melting Pt: 148…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001405
CAS Number: 3859-41-4
EINECS: 223-372-8

Order Now

Specification Test Results

Appearance Ochre to brown Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 153 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR