Trimethyl orthobenzoate 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Trimethyl orthobeznoate, (Trimethoxymethyl)benzene

172920100 172920500
ACRO172920100EA 33.2 GBP
ACRO172920100 ACRO172920500
Trimethyl orthobenzoate 97%
Trimethyl orthobenzoate
Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Boiling Pt: 87…88 °C (7 mmHg)
Flash Pt: 82 °C
MDL Number: MFCD00008474
CAS Number: 707-07-3
EINECS: 211-897-5

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4910 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR