1-Butylboronic acid 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

172380010 172380050 172380250
ACRO172380010EA 18.3 GBP
ACRO172380010 ACRO172380050 ACRO172380250
1-Butylboronic acid 98%
1-Butylboronic acid
Formula: C₄H₁₁BO₂
Melting Pt: 94…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002106
CAS Number: 4426-47-5
EINECS: 224-607-7

Order Now

Specification Test Results

Appearance White to off-white Glistening powder or flakes
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR