4-Nitrophenyl chloroformate 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

170800050 170800250 170801000 170805000
ACRO170800050EA 27.5 GBP
ACRO170800050 ACRO170800250 ACRO170801000 ACRO170805000
4-Nitrophenyl chloroformate 97%
4-Nitrophenyl chloroformate
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 159…162 °C (19 mmHg)
Melting Pt: 73…81 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007321
CAS Number: 7693-46-1
EINECS: 231-706-9
UN: 2923
ADR: 8 II

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 77 to 82 °C
Titration Argentometric ≥96.0 % (ex Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR