Triethanolamine hydrochloride 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

170051000 170055000 170050025
ACRO170051000EA 29.4 GBP
ACRO170051000 ACRO170055000 ACRO170050025
Triethanolamine hydrochloride 99+%
Triethanolamine hydrochloride
Formula: C₆H₁₆ClNO₃
Melting Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00012596
CAS Number: 637-39-8
EINECS: 211-284-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
Loss on drying ≤1.0 % (105°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR