Potassium oxonate 97.5%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Potassium Allantoxanate, Oxonic acid Potassium Salt, Potassium 4,6-Dihydroxy-1,3,5-triazine-2-carboxylate, Allantoxanic acid Potassium Salt, Potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate

168530250 168531000
ACRO168530250EA 74.1 GBP
ACRO168530250 ACRO168531000
Potassium oxonate 97.5%
Potassium oxonate

Appearance: White Crystalline powder

Formula: C₄H₃N₃O₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00010565
CAS Number: 2207-75-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR