Methanol 99.9% for spectroscopy

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

167830025 167835000
ACRO167830025EA 72.1 GBP
ACRO167830025 ACRO167835000
Methanol 99.9% for spectroscopy
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.7 °C (760 mmHg)
Melting Pt: –98 °C
Density: 0.791 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
GC ≥99.85 %
UV at 260 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR