Chloroform 99+% stabilised for spectroscopy

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilisation: Stabilised with amylene

167730025 167735000 167730010 167730250
ACRO167730025EA 113 GBP
ACRO167730025 ACRO167735000 ACRO167730010 ACRO167730250
Chloroform 99+% stabilised for spectroscopy
Chloroform
Formula: CHCl₃
Boiling Pt: 61 °C (760 mmHg)
Melting Pt: –63 °C
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.0 %
UV at 280 nm A: ≤0.005
Color scale ≤10 APHA
Stabilizer 70 to 120 ppm Amylene
Residue after evaporation ≤0.0005 %
Acidity/alkalinity ≤0.0002 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR