Carbon disulphide 99.9% for spectroscopy

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

167710250 167715000 167710025
ACRO167710250EA 27.3 GBP
ACRO167710250 ACRO167715000 ACRO167710025
Carbon disulphide 99.9% for spectroscopy
Carbon disulphide
Formula: CS₂
Boiling Pt: 46 °C (760 mmHg)
Melting Pt: –111 °C
Density: 1.262 g/cm³
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 %
GC ≥99.85 %
UV at 380 nm A: ≤1.3
UV at 390 nm A: ≤0.5
UV at 400 nm A: ≤0.1
UV at 410 nm A: ≤0.05
UV at 420 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR