Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

166250250 166250500 166251000 166252500 166255000
ACRO166250250EA 27.8 GBP
ACRO166250250 ACRO166250500 ACRO166251000 ACRO166252500 ACRO166255000
Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Chloroform-[D1]
Formula: CDCl₃
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.75 %
HPLC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR