4-Nitro-o-toluidine 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Nitro-2-methylaniline, 2-Methyl-4-nitroaniline, 2-Amino-5-nitrotoluene

163950050
ACRO163950050EA 14.6 GBP
ACRO163950050
4-Nitro-o-toluidine 99%
4-Nitro-o-toluidine
Formula: C₇H₈N₂O₂
Melting Pt: 129…133 °C
Density: 1,366 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007734
CAS Number: 99-52-5
EINECS: 202-762-1
UN: 2660
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to brown to khaki Crystalline powder or needles
Infrared spectrum Conforms
Melting point 129 to 133 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR