Bisacrylamide 96%, extra pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

163801000 163805000 163800025
ACRO163801000EA 24.2 GBP
ACRO163801000 ACRO163805000 ACRO163800025
Bisacrylamide 96%, extra pure
Bisacrylamide
Formula: (H₂C=CHCONH)₂CH₂
Boiling Pt: 125 °C (3,33 hPa)
Melting Pt: 185 °C (dec.)
Density: 1,235 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
HPLC ≥95.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR