Veratrole 99+%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

161110010 161111000 161115000
ACRO161110010EA 73.8 GBP
ACRO161110010 ACRO161111000 ACRO161115000
Veratrole 99+%
Veratrole
Formula: C₈H₁₀O₂
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.08 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00008357
CAS Number: 91-16-7
EINECS: 202-045-3
Merck Index: 12,10089

Order Now

Specification Test Results

Appearance Clear liquid or solid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5360 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR