N,N,N-Trimethylanilinium tribromide 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide

160940250 160941000
ACRO160940250EA 31.1 GBP
ACRO160940250 ACRO160941000
N,N,N-Trimethylanilinium tribromide 97%
N,N,N-Trimethylanilinium tribromide
Formula: C₉H₁₄Br₃N
MW: 375.93 g/mol
Melting Pt: 113…117 °C
MDL Number: MFCD00011789
CAS Number: 4207-56-1
EINECS: 224-127-8
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 117 °C
Titration ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR