1-Pentanol 99%, pure

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Pentyl alcohol, 1-Hydroxy pentane, n-Amyl alcohol, Amyl alcohol, Butyl carbinol

160600010 160600025 160600250 160600251
ACRO160600010EA 34 GBP
ACRO160600010 ACRO160600025 ACRO160600250 ACRO160600251
1-Pentanol 99%, pure
1-Pentanol
Formula: H₃C(CH₂)₃CH₂OH
MW: 88.15 g/mol
Boiling Pt: 137…139 °C (760 mmHg)
Melting Pt: –78 °C
Density: 0.811 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00002977
CAS Number: 71-41-0
EINECS: 200-752-1
UN: 1105
ADR: 3 III
Merck Index: 13,07195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92dac1192eb8e068fc1804fdcf488129a2385918&vid=d4c7f2c5672a97c6dfaddc50807d768068096252&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4110 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR