Hydrindantin dihydrate 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

159280100 159280500 159282500
ACRO159280100EA 35.5 GBP
ACRO159280100 ACRO159280500 ACRO159282500
Hydrindantin dihydrate 96%
Hydrindantin dihydrate
Formula: C₁₈H₁₀O₆·2H₂O
Melting Pt: >240 °C
Storage Temperature: Ambient
MDL Number: MFCD00149242
CAS Number: 5950-69-6
EINECS: 227-713-1

Order Now

Specification Test Results

Appearance Light beige to pink-beige Powder
Infrared spectrum Conforms
Loss on drying 9 % to 11 % (105°C, 3 hr)
HPLC ≥95 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.003 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.0005 %
Lead (Pb) ≤0.001 %
Sodium (Na) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR