Maleimide 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

156710050 156710100 156710250 156711000 156715000
ACRO156710050EA 42.5 GBP
ACRO156710050 ACRO156710100 ACRO156710250 ACRO156711000 ACRO156715000
Maleimide 98%
Maleimide
Formula: C₄H₃NO₂
Boiling Pt: 102 - 105 °C (55 -rr)
Melting Pt: 90…94 °C
Density: 1,2493 g/cm³ (106 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005494
CAS Number: 541-59-3
EINECS: 208-787-4
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
GC ≥97.5 %
Titration with NaOH ≥97.5 %
Substance darkened by H2SO4 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR